Sodium acetate, anhydrous 99+% ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Acetic acid sodium salt

AC42425-0025 AC42425-5000
TS42425-0025EA 259 USD
TS42425-0025 TS42425-5000
Sodium acetate, anhydrous 99+% ACS
Sodium acetate
Formula: H₃CCOONa
MW: 82.03 g/mol
Boiling Pt: 881.4
Melting Pt: 324
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1.0 % (120°C)
pH 7.0 to 9.2 (5 % at 25.0°C)
Titration with HClO4 ≥99.0 %
Chloride (Cl) ≤0.002 %
Phosphate (PO4) ≤0.001 %
Sulfate (SO4) ≤0.003 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.002 %
Heavy metals (ICP-OES) ≤0.001 %
Insoluble matter ≤0.01 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR