Chromium(III) nitrate nonahydrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Chromium trinitrate nonahydrate, Chromic nitrate nonahydrate, Chromium nitrate nonahydrate
AC21920-0010
AC21920-0051
AC21920-1000
AC21920-5000
TS21920-0010EA
274
USD
TS21920-0010
TS21920-0051
TS21920-1000
TS21920-5000
Chromium(III) nitrate nonahydrate 99%
Chromium (III) nitrate nonahydrate
Formula:
CrN₃O₉·9H₂O MW: 400.15 g/mol Melting Pt: 60 Storage Temperature: Ambient |
MDL Number:
MFCD00149659 CAS Number: 7789-02-8 EINECS: 236-921-1 UN: 2720 ADR: 5.1 III Merck Index: 13,02246 |
Specification Test Results
Appearance | Dark purple to black Crystalline chunks |
Titration Iodometric | ≥98.5 % |
pH | 2 to 3 (5 % in water) |
Chloride (Cl) | ≤20 ppm |
Sulfate (SO4) | ≤50 ppm |
Ammonium (NH4) | ≤10 ppm |
Cadmium (Cd) | ≤10 ppm |
Calcium (Ca) | ≤50 ppm |
Cobalt (Co) | ≤10 ppm |
Copper (Cu) | ≤10 ppm |
Iron (Fe) | ≤200 ppm |
Lead (Pb) | ≤20 ppm |
Magnesium (Mg) | ≤50 ppm |
Manganese (Mn) | ≤10 ppm |
Nickel (Ni) | ≤50 ppm |
Sodium (Na) | ≤50 ppm |
Strontium (Sr) | ≤50 ppm |
Zinc (Zn) | ≤10 ppm |
Subst not pptd by NH4OH | ≤0.2 % (as SO4) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...