Calcium fluoride, anhydrous 99%, pure

Supplier: Thermo Scientific Chemicals

AC21825-0100 AC21825-1000 AC21825-5000
TS21825-0100EA 24 USD
TS21825-0100 TS21825-1000 TS21825-5000
Calcium fluoride, anhydrous 99%, pure
Calcium fluoride
Formula: CaF₂
MW: 78.07 g/mol
Boiling Pt: 2500
Melting Pt: 1418
Density: 3.18 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010907
CAS Number: 7789-75-5
EINECS: 232-188-7
Merck Index: 13,01669

Order Now

Specification Test Results

Appearance White to off-white Powder
Assay ≥98.5 % (CaF2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR