Bromotrimethylsilane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC18665-0050
AC18665-0250
AC18665-1000
AC18665-5000
TS18665-0050EA
55
USD
TS18665-0050
TS18665-0250
TS18665-1000
TS18665-5000
Bromotrimethylsilane 98%
Bromotrimethylsilane
Formula:
C₃H₉BrSi Boiling Pt: 79 °C (760 mmHg) Melting Pt: –43 °C Density: 1.16 g/cm³ Flash Pt: 32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000048 CAS Number: 2857-97-8 EINECS: 220-672-0 UN: 2920 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4200 to 1.4250 (20°C, 589 nm) |
Titration Argentometric | ≥97.5 % |
Titration with NaOH | 97.5 to 102.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...