Bismuth(III) oxide 99.9%, extra pure

Supplier: Thermo Scientific Chemicals

Synonyms: Bismuth Trioxide, Dibismuth trioxide

AC20807-0025 AC20807-1000 AC20807-5000
TS20807-0025EA 777 USD
TS20807-0025 TS20807-1000 TS20807-5000
Bismuth(III) oxide 99.9%, extra pure
Bismuth (III) oxide
Formula: BiO₃
MW: 465.96 g/mol
Boiling Pt: 1890
Melting Pt: 825
Density: 8.9 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003462
CAS Number: 1304-76-3
EINECS: 215-134-7
Merck Index: 13,01273

Order Now

Specification Test Results

Appearance Yellow Powder
Loss on drying ≤0.2 %
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤10 ppm
Bismuth (Bi) 89.5 to 89.9 %
Sodium (Na) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR