tri-Sodium citrate dihydrate 99%, pure
Supplier: Thermo Scientific Chemicals
AC22713-0010
AC22713-0051
AC22713-0250
AC22713-5000
TS22713-0010EA
66
USD
TS22713-0010
TS22713-0051
TS22713-0250
TS22713-5000
tri-Sodium citrate dihydrate 99%, pure
tri-Sodium citrate dihydrate
Formula:
Na₃C₆H₅O₇·2H₂O Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150031 CAS Number: 6132-04-3 EINECS: 200-675-3 Merck Index: 13,08675 |
|
Specification Test Results
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Water | 11 to 13 % |
Titration with HClO4 | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...