Potassium hexacyanoferrate(III) 98%, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide

AC19678-0010 AC19678-0050 AC19678-1000 AC19678-5000
TS19678-0010EA 116 USD
TS19678-0010 TS19678-0050 TS19678-1000 TS19678-5000
Potassium hexacyanoferrate(III) 98%, pure
Potassium hexacyanoferrate (III)
Formula: K₃Fe(CN)₆
MW: 329.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011392
CAS Number: 13746-66-2
EINECS: 237-323-3
Merck Index: 13,07712

Order Now

Specification Test Results

Appearance Orange to red Crystalline powder or crystals
Titration Iodometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR