Triethyl citrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Triethyl 2-hydroxypropane-1,2,3-tricarboxylate, Hydroxytricarballylic acid triethyl ester, Citric acid triethyl ester

AC37506-0010 AC37506-0025 AC37506-2500
TS37506-0010EA 105 USD
TS37506-0010 TS37506-0025 TS37506-2500
Triethyl citrate 99%
Triethyl citrate
Formula: C₁₂H₂₀O₇
Boiling Pt: 294 °C (760 mmHg)
Melting Pt: –46 °C
Density: 1.136 g/cm³
Flash Pt: 151 °C
Storage Temperature: Ambient
MDL Number: MFCD00009201
CAS Number: 77-93-0
EINECS: 201-070-7

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4430 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤50 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR