Copper(I) cyanide 99%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Copper cyanide, Cuprous cyanide

AC20208-0010 AC20208-0051 AC20208-2500
TS20208-0010EA 209 USD
TS20208-0010 TS20208-0051 TS20208-2500
Copper(I) cyanide 99%, extra pure
Copper (I) cyanide
Formula: CCuN
MW: 89.56 g/mol
Melting Pt: 474
Density: 2.92 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010975
CAS Number: 544-92-3
EINECS: 208-883-6
UN: 1587
ADR: 6.1 II
Merck Index: 13,02691

Order Now

Specification Test Results

Appearance White to beige to green Powder
Assay ≥98.5 % (ex Cu)
Arsenic (As) ≤10 ppm
Iron (Fe) ≤100 ppm
Lead (Pb) ≤20 ppm
Tin (Sn) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR