Potassium perchlorate ACS

Supplier: Thermo Scientific Chemicals
Danger

AC22137-1000 AC22137-5000
TS22137-1000EA 53 USD
TS22137-1000 TS22137-5000
Potassium perchlorate ACS
Potassium perchlorate
Formula: ClKO₄
MW: 138.55 g/mol
Melting Pt: 400 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011362
CAS Number: 7778-74-7
UN: 1489
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration 99.0 to 100.5 %
Chloride (Cl) ≤0.003 %
Sulfate (SO4) ≤0.001 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤5 ppm
Sodium (Na) ≤0.02 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR