Chromium(III) chloride, anhydrous 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Chromic chloride, Chromium trichloride
AC19047-0050
AC19047-0250
AC19047-1000
TS19047-0050EA
38
USD
TS19047-0050
TS19047-0250
TS19047-1000
Chromium(III) chloride, anhydrous 99%
Chromium (III) chloride
Formula:
Cl₃Cr MW: 158.35 g/mol Melting Pt: 1152 Density: 2.8 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00010948 CAS Number: 10025-73-7 UN: 3260 ADR: 8 III Merck Index: 12,02278 |
Specification Test Results
Appearance | Purple Glistening powder or flakes |
Assay | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...