Hexabutylditin 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hexa-n-butylditin, Hexabutyldistannane, Bis(tributyltin)
AC35151-0100
AC35151-0500
TS35151-0100EA
68
USD
TS35151-0100
TS35151-0500
Hexabutylditin 98%
Hexabutylditin
Formula:
C₂₄H₅₄Sn₂ Boiling Pt: 197…198 °C (10 mmHg) Density: 1.148 g/cm³ Flash Pt: >110 °C Storage Temperature: Freezer |
MDL Number:
MFCD00009417 CAS Number: 813-19-4 EINECS: 212-383-3 UN: 2788 ADR: 6.1,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5100 to 1.5130 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...