Cobalt 99+%, powder, pure

Supplier: Thermo Scientific Chemicals
Danger

AC45391-0010 AC45391-2500
TS45391-0010EA 558 USD
TS45391-0010 TS45391-2500
Cobalt 99+%, powder, pure
Cobalt
Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
Merck Index: 12,02488

Order Now

Specification Test Results

Appearance Gray Powder
Titration Complexometric ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR