Ammonium iron (II) sulfate hexahydrate 98%, Extra Pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ferrous ammonium sulfate hexahydrate
, Ammonium iron(Ⅱ) sulfate hexahydrate
, Ammonium ferrous sulphate hexahydrate
, di-Ammonium iron (II) sulphate hexahydrate
, Ammonium ferrous sulfate hexahydrate
, Ferrous ammonium sulphate hexahydrate
, Ammonium iron(II) sulphate hydrate
, di-Ammonium iron (II) sulfate hexahydrate
, Ammonium iron(Ⅱ) sulphate hexahydrate
, Ammonium iron(II) sulfate hydrate
AC19325-0050
TS19325-0050EA
152
USD
TS19325-0050
Ammonium iron (II) sulfate hexahydrate 98%, Extra Pure
Ammonium iron (II) sulphate hexahydrate
Formula:
(NH₄)₂Fe(SO₄)₂·6H₂O MW: 392.14 g/mol Melting Pt: 100 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00150530 CAS Number: 7783-85-9 |
Specification Test Results
Appearance | Green to blue-green to blue to dark blue Powder |
pH | 3 to 5 |
Titration with KMnO4 | ≥97.5 % |
Insoluble matter | ≤0.05 % (in dilute H2SO4) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...