Ammonium iron (II) sulfate hexahydrate 99% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: di-Ammonium iron (II) sulfate hexahydrate, Ferrous Ammonium sulphate Hexahydrate, Ferrous ammonium sulfate hexahydrate, Ferrous Ammonium sulfate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium iron(Ⅱ) sulfate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate, Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate hydrate

AC42372-0030 AC42372-1000 AC42372-5000
TS42372-0030EA 598 USD
TS42372-0030 TS42372-1000 TS42372-5000
Ammonium iron (II) sulfate hexahydrate 99% ACS
Ammonium iron (II) sulphate hexahydrate
Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
MW: 392.14 g/mol
Melting Pt: 100 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150530
CAS Number: 7783-85-9
EINECS: 233-151-8

Order Now

Specification Test Results

Appearance Blue-green Crystals
Titration with KMnO4 98.5 to 101.5 %
Phosphate (PO4) ≤0.003 %
Calcium (Ca) ≤0.005 %
Copper (Cu) ≤0.003 %
Magnesium (Mg) ≤0.002 %
Manganese (Mn) ≤0.01 %
Potassium (K) ≤0.002 %
Sodium (Na) ≤0.02 %
Zinc (Zn) ≤0.003 %
Ferric iron (Fe 3+) ≤100 ppm (May increase due to inherent oxidation on storage)
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR