Ammonium persulfate (APS) 99+% for molecular biology DNase-, RNase-, protease-free
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Peroxidisulfuric acid diammonium salt, Ammonium persulfate, Peroxidisulphuric acid diammonium salt, Ammonium persulphate
AC32708-1000
AC32708-5000
TS32708-1000EA
58
USD
TS32708-1000
TS32708-5000
Ammonium persulfate (APS) 99+% for molecular biology DNase-, RNase-, protease-free
Ammonium peroxodisulphate (APS)
Formula:
(NH₄)₂S₂O₈ Melting Pt: 120 °C (dec.) Density: 1.98 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003390 CAS Number: 7727-54-0 EINECS: 231-786-5 UN: 1444 ADR: 5.1,III Merck Index: 13,00545 |
|
Specification Test Results
Appearance | White Powder |
Titration Iodometric | ≥99 % |
Acidity | ≤0.2 % (H2SO4) |
Dnase,Rnase,Protease activity | none detected |
Chloride (Cl) | ≤20 ppm |
Iron (Fe) | ≤10 ppm |
Heavy metals (as Pb) | ≤50 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...