Diammonium hydrogen orthophosphate 99+% for analysis

Supplier: Thermo Scientific Chemicals

Synonyms: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate

AC20182-0025 AC20182-2500 AC20182-5000
TS20182-0025EA 406 USD
TS20182-0025 TS20182-2500 TS20182-5000
Diammonium hydrogen orthophosphate 99+% for analysis
di-Ammonium hydrogen orthophosphate
Formula: (NH₄)₂HPO₄
MW: 132.06 g/mol
Melting Pt: 185
Storage Temperature: Ambient
MDL Number: MFCD00010891
CAS Number: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Merck Index: 13,00546

Order Now

Specification Test Results

Appearance White Adhering crystals
pH 7.9 to 8.3 (5 % at 20°C)
Titration with HCl ≥99.0 %
Chloride (Cl) ≤5 ppm
Nitrate (NO3) ≤100 ppm
Sulfate (SO4) ≤200 ppm
Iron (Fe) ≤10 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤100 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR