Dipotassium hydrogen orthophosphate trihydrate 99+% for analysis

Supplier: Thermo Scientific Chemicals

AC20593-0025 AC20593-5000
TS20593-0025EA 494 USD
TS20593-0025 TS20593-5000
Dipotassium hydrogen orthophosphate trihydrate 99+% for analysis
di-Potassium hydrogen orthophosphate trihydrate
Formula: K₂HPO₄·3H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149926
CAS Number: 16788-57-1
EINECS: 231-834-5

Order Now

Specification Test Results

Appearance White Adhering crystals
pH 8.5 to 9.6 (5 % solution)
Titration with HClO4 ≥99.0 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤100 ppm
Arsenic (As) ≤1 ppm
Copper (Cu) ≤30 ppm
Iron (Fe) ≤30 ppm
Lead (Pb) ≤30 ppm
Nickel (Ni) ≤30 ppm
Insoluble matter ≤0.01 % (in water)
Nitrogen compounds (as N) ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR