Diammonium hydrogen citrate 98%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ammonium citrate dibasicDiammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylateCitric acid diammonium salt

AC39250-5000 AC39250-0025 AC39250-1000
TS39250-0025EA 186 USD
TS39250-0025 TS39250-1000 TS39250-5000
Diammonium hydrogen citrate 98%, pure
di-Ammonium hydrogen citrate
Formula: HOC(CO₂H)(CH₂CO₂NH₄)₂
MW: 226.19 g/mol
Density: 1.5 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00013068
CAS Number: 3012-65-5

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR