Potassium citrate monohydrate 99%, extra pure

Supplier: Thermo Scientific Chemicals

Synonyms: Potassium citrate tribasic monohydrate, Citric acid tripotassium salt monohydrate, Tripotassium citrate monohydrate, Tripotassium 2-hydroxypropane-1,2,3-tricarboxylate monohydrate

AC24421-0010 AC24421-0050
TS24421-0010EA 82 USD
TS24421-0010 TS24421-0050
Potassium citrate monohydrate 99%, extra pure
Potassium citrate monohydrate
Formula: HOC(COOK)(CH₂COOK)₂·H₂O
MW: 324.41 g/mol
Melting Pt: 230
Storage Temperature: Ambient
MDL Number: MFCD00150442
CAS Number: 6100-05-6
Merck Index: 13,07706
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc3e6bae942870a760ea6cb418203db7458ff3dc&vid=2c2580a7bfc83e60c4dfca9f8af4ac3b3db56b27&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to white Powder or crystals
Infrared spectrum Conforms
Water 4 to 6 %
Acidity/alkalinity Passes test
Titration with HClO4 ≥98.5 %
Heavy metals (as Pb) ≤0.001 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR