Ammonium carbonate for analysis
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Salt of hartshorn
AC19665-0010
AC19665-0025
AC19665-1000
AC19665-5000
TS19665-0010EA
103
USD
TS19665-0010
TS19665-0025
TS19665-1000
TS19665-5000
Ammonium carbonate for analysis
Ammonium carbonate
Formula:
(NH₄)₂CO₃ Melting Pt: 58 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00010890 CAS Number: 506-87-6 EINECS: 208-058-0 Merck Index: 13,00508 |
Specification Test Results
Appearance | White Powder or crystals and/or chunks |
Non-volatile matter | ≤0.01 % |
Titration with HCl | ≥30.0 % (NH3) |
Chloride (Cl) | ≤20 ppm |
Sulfate (SO4) | ≤50 ppm |
Iron (Fe) | ≤10 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...