Cesium carbonate 99.5% analysis_for
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC19204-0025
AC19204-0050
AC19204-0250
AC19204-1000
AC19204-5000
TS19204-0025EA
2162
USD
TS19204-0025
TS19204-0050
TS19204-0250
TS19204-1000
TS19204-5000
Cesium carbonate 99.5% analysis_for
Cesium carbonate
Formula:
Cs₂CO₃ MW: 325.82 g/mol Melting Pt: 610 Storage Temperature: Ambient |
MDL Number:
MFCD00010957 CAS Number: 534-17-8 EINECS: 208-591-9 Merck Index: 13,02020 |
Specification Test Results
Appearance | White Crystalline powder |
Assay | ≥99.4 % (By difference) |
Loss on drying | ≤0.5 % (500°C) |
Chloride (Cl) | ≤300 ppm |
Sulfate (SO4) | ≤500 ppm |
Aluminium (Al) | ≤100 ppm |
Barium (Ba) | ≤100 ppm |
Calcium (Ca) | ≤20 ppm |
Potassium (K) | ≤400 ppm |
Rubidium (Rb) | ≤3000 ppm |
Sodium (Na) | ≤300 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...