Niobium(V) chloride 99.8%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Niobium pentachloride

AC20169-0100 AC20169-0500
TS20169-0100EA 47 USD
TS20169-0100 TS20169-0500
Niobium(V) chloride 99.8%
Niobium (V) chloride
Formula: Cl₅Nb
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 204.7…209.5 °C
MDL Number: MFCD00011127
CAS Number: 10026-12-7
EINECS: 233-059-8
UN: 3260
ADR: 8,II
Merck Index: 12,06649

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Assay ≥99.75 %
Aluminium (Al) ≤4 ppm
Bismuth (Bi) ≤0.5 ppm
Boron (B) ≤2 ppm
Calcium (Ca) ≤3 ppm
Chromium (Cr) ≤0.5 ppm
Cobalt (Co) ≤0.2 ppm
Copper (Cu) ≤0.5 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤1 ppm
Magnesium (Mg) ≤1 ppm
Manganese (Mn) ≤0.5 ppm
Molybdenum (Mo) ≤3 ppm
Nickel (Ni) ≤0.5 ppm
Potassium (K) ≤1 ppm
Silicon (Si) ≤3 ppm
Sodium (Na) ≤1 ppm
Tantalium (Ta) ≤25 ppm
Tin (Sn) ≤1 ppm
Titanium (Ti) ≤3 ppm
Tungsten (W) ≤2 ppm
Vanadium (V) ≤2 ppm
Zirconium (Zr) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR