Tetrabutylammonium tribromide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC12397-0250
AC12397-1000
AC12397-5000
TS12397-0250EA
63
USD
TS12397-0250
TS12397-1000
TS12397-5000
Tetrabutylammonium tribromide 98+%
Tetrabutylammonium tribromide
Formula:
C₁₆H₃₆Br₃N Melting Pt: 71…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012110 CAS Number: 38932-80-8 |
Specification Test Results
Appearance | Orange Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 71 to 75 °C |
Titration Iodometric | ≥98.0 % |
Specific optical rotation | -22° to -26° (20 °C, 589 nm) (c=1, ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...