Silver(I) oxide 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
di-Silver oxide, Disilver oxide, Silver oxide
AC20580-0100
AC20580-0500
AC20580-2500
TS20580-0100EA
78
USD
TS20580-0100
TS20580-0500
TS20580-2500
Silver(I) oxide 99+%
Silver(I) oxide
Formula:
Ag₂O Melting Pt: 800…830 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003404 CAS Number: 20667-12-3 EINECS: 243-957-1 UN: 1479 ADR: 5.1,I Merck Index: 12,08665 |
Specification Test Results
Appearance | Black Powder |
Elemental Analysis | ≥99.0 % |
Nitrate (NO3) | ≤50 ppm |
Sulfate (SO4) | ≤100 ppm |
Aluminium (Al) | ≤10 ppm |
Iron (Fe) | ≤30 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...