Xenon difluoride 99.5+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Xenon(II) fluoride

AC27855-0010 AC27855-0050
TS27855-0010EA 116 USD
TS27855-0010 TS27855-0050
Xenon difluoride 99.5+%
Xenon difluoride
Formula: F₂Xe
Melting Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD00040538
CAS Number: 13709-36-9
EINECS: 237-251-2
UN: 3085
ADR: 5.1,II
Merck Index: 14,10072

Order Now

Specification Test Results

Appearance White Crystals
Purity ≥99.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR