Selenium dioxide 99.999% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

AC19398-0100 AC19398-0500
TS19398-0100EA 131 USD
TS19398-0100 TS19398-0500
Selenium dioxide 99.999% (trace metals basis)
Selenium dioxide
Formula: SeO₂
Melting Pt: 315 °C
Storage Temperature: Ambient
MDL Number: MFCD00003562
CAS Number: 7446-08-4
EINECS: 231-194-7
UN: 3283
ADR: 6.1,II
Merck Index: 13,08508

Order Now

Specification Test Results

Appearance Off-white Crystalline powder
Total trace metal impurities ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR