Phosphorus pentasulfide 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
di-Phosphorus pentasulfide, di-Phosphorus pentasulphide
AC19672-0010
AC19672-5000
TS19672-0010EA
90
USD
TS19672-0010
TS19672-5000
Phosphorus pentasulfide 98+%
Phosphorus pentasulphide
Formula:
P₂S₅ Boiling Pt: 514 °C (760 mmHg) Melting Pt: 282…286 °C Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011441 CAS Number: 1314-80-3 EINECS: 215-242-4 UN: 1340 ADR: 4.3,II Merck Index: 12,07511 |
Specification Test Results
Appearance | Yellow to green Powder |
Assay | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...