Phosphoryl tribromide 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Phosphorus(V) oxybromide, Phosphorus (V) oxide bromide, Phosphoryl bromide, Phosphorus oxybromide
AC38845-0100
AC38845-0250
AC38845-1000
TS38845-0100EA
70
USD
TS38845-0100
TS38845-0250
TS38845-1000
Phosphoryl tribromide 95%
Phosphoryl tribromide
Formula:
Br₃OP Boiling Pt: 192 °C (760 mmHg) Melting Pt: 56 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00036298 CAS Number: 7789-59-5 EINECS: 232-177-7 UN: 1939 ADR: 8,II Merck Index: 12,07505 |
Specification Test Results
Appearance | Colorless to gray-brown Crystals or fused mass |
Titration Argentometric | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...