Barium nitrate, pure

Supplier: Thermo Scientific Chemicals
Danger

AC45380-0025 AC45380-0100 AC45380-5000
TS45380-0025EA 102 USD
TS45380-0025 TS45380-0100 TS45380-5000
Barium nitrate, pure
Barium nitrate
Formula: Ba(NO₃)₂
Melting Pt: 592 °C
Storage Temperature: Ambient
MDL Number: MFCD00003442
CAS Number: 10022-31-8
UN: 1446
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Complexometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR