Magnesium chloride, anhydrous 99.9% (trace metals basis), pure
Supplier: Thermo Scientific Chemicals
AC19706-0500
AC19706-0100
TS19706-0500EA
348
USD
TS19706-0500
TS19706-0100
Magnesium chloride, anhydrous 99.9% (trace metals basis), pure
Magnesium chloride
Formula:
MgCl₂ MW: 95.21 g/mol Boiling Pt: 1412 °C (760 mmHg) Melting Pt: 708 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011106 CAS Number: 7786-30-3 |
Specification Test Results
Appearance | White Powder to beads |
Total trace metal impurities | ≤0.1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...