Sodium sulfite, anhydrous 97%, extra pure

Supplier: Thermo Scientific Chemicals

AC19663-0010 AC19663-0025 AC19663-0100
TS19663-0010EA 72 USD
TS19663-0010 TS19663-0025 TS19663-0100
Sodium sulfite, anhydrous 97%, extra pure
Sodium sulphite
Formula: Na₂SO₃
MW: 126.04 g/mol
Melting Pt: >500 °C (dec.)
Density: 2.63 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 13,08757

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Titration Iodimetric ≥96.0 %
pH 8.5 to 10.5 (5 % solution)
Sodium chloride (NaCl) ≤50 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR