Disodium sulfide 90+%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium sulfide, Sodium sulphide
AC31018-0100
AC31018-2500
TS31018-0100EA
162
USD
TS31018-0100
TS31018-2500
Disodium sulfide 90+%, pure
Disodium sulphide
Formula:
Na₂S MW: 78.05 g/mol Boiling Pt: 174 Storage Temperature: Refrigerator |
MDL Number:
MFCD00003498 CAS Number: 1313-82-2 EINECS: 215-211-5 UN: 1385 ADR: 4.2 II REACH: 01-2119513694-38 Merck Index: 12,08830 |
Specification Test Results
Appearance | Yellow to beige Powder and/or lumps |
Titration Iodimetric | ≥90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...