Sodium dihydrogen phosphate, anhydrous 98%, Extra Pure

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic, Sodium dihydrogen phosphate

AC44817-0010 AC44817-0050
TS44817-0010EA 71 USD
TS44817-0010 TS44817-0050
Sodium dihydrogen phosphate, anhydrous 98%, Extra Pure
Sodium dihydrogen phosphate
Formula: NaH₂PO₄
MW: 119.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003527
CAS Number: 7558-80-7

Order Now

Specification Test Results

Appearance White Crystalline powder
Water ≤2 %
Loss on drying ≤1 % (130°C)
pH 4.1 to 4.5
Titration with HClO4 97.5 to 103 %
Heavy metals (as Pb) ≤0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR