penta-Sodium triphosphate for analysis

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium triphosphate pentabasic, Sodium tripolyphosphate

AC39396-0250 AC39396-1000 AC39396-5000
TS39396-0250EA 34 USD
TS39396-0250 TS39396-1000 TS39396-5000
penta-Sodium triphosphate for analysis
Pentasodium triphosphate
Formula: Na₅O₁₀P₃
Melting Pt: 622 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003514
CAS Number: 7758-29-4

Order Now

Specification Test Results

Appearance White Powder
pH 9.0 to 10.0 (1% solution)
Titration ≥98.0 %
Cadmium (Cd) ≤50 ppm
Calcium (Ca) ≤200 ppm
Copper (Cu) ≤50 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm
Nickel (Ni) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR