Sodium peroxide ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC31576-0250
AC31576-1000
AC31576-5000
TS31576-0250EA
73
USD
TS31576-0250
TS31576-1000
TS31576-5000
Sodium peroxide ACS
Sodium peroxide
Formula:
Na₂O₂ Melting Pt: 460 °C (dec.) Density: 2.8 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003497 CAS Number: 1313-60-6 EINECS: 215-209-4 UN: 1504 ADR: 5.1,I Merck Index: 13,08728 |
Specification Test Results
Appearance | Yellow Powder or granules (May contain a small quantity of discolored material from the manufacturing process) |
Titration with KMnO4 | ≥93.0 % |
Chloride (Cl) | ≤20 ppm |
Phosphate (PO4) | ≤5 ppm |
Sulfate (SO4) | ≤10 ppm |
Iron (Fe) | ≤50 ppm |
Heavy metals (as Pb) | ≤20 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...