Potassium iodate 98%, extra pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC19674-0025
AC19674-1000
AC19674-5000
TS19674-0025EA
740
USD
TS19674-0025
TS19674-1000
TS19674-5000
Potassium iodate 98%, extra pure
Potassium iodate
Formula:
KlO₃ Melting Pt: 560 °C Density: 3.93 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011406 CAS Number: 7758-05-6 EINECS: 231-831-9 UN: 1479 ADR: 5.1,II Merck Index: 13,07726 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Titration Iodometric | ≥97.5 % |
Loss on drying | ≤0.5 % (105°C, 3 hrs) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...