Potassium iodide 99.995% (trace metals basis), Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dipotassium diiodideKali iodide

AC19379-2500 AC19379-0100 AC19379-0500
TS19379-0100EA 69 USD
TS19379-0100 TS19379-2500 TS19379-0500
Potassium iodide 99.995% (trace metals basis), Extra Pure
Potassium iodide
Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330 °C (760 mmHg)
Melting Pt: 680 °C
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 12,07809

Order Now

Specification Test Results

Appearance White Powder or crystals
Total trace metal impurities ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR