Sodium fluoride 99+% for analysis

Supplier: Thermo Scientific Chemicals
Danger

AC20129-0250 AC20129-5000
TS20129-0250EA 32 USD
TS20129-0250 TS20129-5000
Sodium fluoride 99+% for analysis
Sodium fluoride
Formula: NaF
Boiling Pt: 1700 °C (1013 hPa)
Melting Pt: 993 °C
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
UN: 1690
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Powder
Loss on drying ≤0.2 % (150°C)
Titration ≥99.0 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤1000 ppm
Sodium fluosilicate ≤0.1 %
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤10 ppm
Free acid (HF) ≤0.1 %
Free alkali (NaOH) ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR