Potassium chloride 99+% ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Potassiumchloride

AC42409-0010 AC42409-0050 AC42409-0250
TS42409-0010EA 139 USD
TS42409-0010 TS42409-0050 TS42409-0250
Potassium chloride 99+% ACS
Potassium chloride
Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420
Melting Pt: 773
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Argentometric 99.0 to 100.5 %
pH 5.4 to 8.6 (5% soln. at 25°C)
Bromide (Br) ≤0.01 %
Iodide (I) ≤20 ppm
Chlorate and Nitrate (as NO3) ≤0.003 %
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤10 ppm
Barium (Ba) Passes test (Limit about 0.001%)
Calcium (Ca) ≤20 ppm
Iron (Fe) ≤3 ppm
Magnesium (Mg) ≤10 ppm
Sodium (Na) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR