Sodium hydrogen carbonate 99+%, extra_pure

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium bicarbonate

AC12336-0010 AC12336-0050 AC12336-0250
TS12336-0010EA 92 USD
TS12336-0010 TS12336-0050 TS12336-0250
Sodium hydrogen carbonate 99+%, extra_pure
Sodium hydrogen carbonate
Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration with HCl ≥99.0 %
Chloride (Cl) ≤100 ppm
Ammonium (NH4) ≤20 ppm
Calcium (Ca) ≤100 ppm
Iron (Fe) ≤10 ppm
Potassium (K) ≤100 ppm
Insoluble matter ≤0.015 %
Total sulfur (S) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR