Potassium hydrogen carbonate, extra pure

Supplier: Thermo Scientific Chemicals

Synonyms: Potassium bicarbonate

AC45064-0010 AC45064-0050 AC45064-0250
TS45064-0010EA 40 USD
TS45064-0010 TS45064-0050 TS45064-0250
Potassium hydrogen carbonate, extra pure
Potassium hydrogen carbonate
Formula: KHCO₃
MW: 100.12 g/mol
Melting Pt: 292
Storage Temperature: Ambient
MDL Number: MFCD00011402
CAS Number: 298-14-6
Merck Index: 13,07691

Order Now

Specification Test Results

Appearance White to off-white Solid
Titration with HCl ≥98.5 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR