Sodium carbonate, anhydrous 99.8%, powder for analysis

Supplier: Thermo Scientific Chemicals
Warning

AC20781-0010 AC20781-0050
TS20781-0010EA 100 USD
TS20781-0010 TS20781-0050
Sodium carbonate, anhydrous 99.8%, powder for analysis
Sodium carbonate
Formula: Na₂CO₃
Boiling Pt: 1600 °C (760 mmHg)
Melting Pt: 851 °C
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline or granular powder
Loss on drying ≤0.5 % (300°C)
Titration with HCl ≥99.8 % (On dried substance)
Chloride (Cl) ≤10 ppm
Phosphate (PO4) ≤10 ppm
Silicate (as SiO2) ≤0.002 %
Aluminium (Al) ≤10 ppm
Arsenic (As) ≤0.5 ppm
Cadmium (Cd) ≤5 ppm
Calcium (Ca) ≤50 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤5 ppm
Lead (Pb) ≤5 ppm
Magnesium (Mg) ≤20 ppm
Potassium (K) ≤50 ppm
Zinc (Zn) ≤5 ppm
Total nitrogen compounds ≤0.001 % (as N)
Total sulfur (as SO4) ≤0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR