Sodium bromate 99+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

AC20887-0025 AC20887-1000 AC20887-5000
TS20887-0025EA 350 USD
TS20887-0025 TS20887-1000 TS20887-5000
Sodium bromate 99+%, Extra Pure
Sodium bromate
Formula: BrNaO₃
Melting Pt: 381 °C (dec.)
Density: 3.33 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003476
CAS Number: 7789-38-0
EINECS: 232-160-4
UN: 1494
ADR: 5.1,II
Merck Index: 12,08736

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Iodometric ≥99.0 %
Bromide (Br) ≤500 ppm
Sulfate (SO4) ≤500 ppm
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR