Methylcyclohexane 99% for spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

AC43377-0010 AC43377-0025
TS43377-0010EA 216 USD
TS43377-0010 TS43377-0025
Methylcyclohexane 99% for spectroscopy
Methylcyclohexane
Formula: C₆H₁₁CH₃
Boiling Pt: 101 °C (760 mmHg)
Melting Pt: –126.3 °C
Density: 0.77 g/cm³
Flash Pt: –3 °C
Storage Temperature: Ambient
MDL Number: MFCD00001497
CAS Number: 108-87-2
UN: 2296
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f7f0d5a1c32138da16df4781897a9c0184e42b7&vid=39a9392c8629d0e2fc79134840e6d8a8dc8a76e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.5 %
GC ≥98.5 %
UV at 260 nm A: ≤0.05
Color scale ≤5 APHA
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR