1-Propanol ACS for spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC43436-0010
AC43436-0025
TS43436-0010EA
96
USD
TS43436-0010
TS43436-0025
1-Propanol ACS for spectroscopy
1-Propanol
Formula:
H₃CCH₂CH₂OH Boiling Pt: 97 °C (760 mmHg) Melting Pt: –127 °C Density: 0.8 g/cm³ Flash Pt: 15 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002941 CAS Number: 71-23-8 UN: 1274 ADR: 3,II |
|
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Water | ≤0.2 % |
Solubility | Clear colorless |
GC | ≥99.5 % |
UV | at 220 nm A: ≤0.7 |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.001 % |
Carbonyl compounds | ≤0.03 % (as C2H5CHO) |
Isopropyl alcohol | ≤0.05 % |
Titrable acid | 0.0004 meq/g |
Ethanol | ≤0.01 % |
Methanol | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...