Europium trinitrate hexahydrate 99.9% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

AC19668-0050
TS19668-0050EA 139 USD
TS19668-0050
Europium trinitrate hexahydrate 99.9% (trace metals basis)
Europium trinitrate hexahydrate
Formula: EuN₃O₉·6H₂O
MW: 446.07 g/mol
Melting Pt: 85 °C
MDL Number: MFCD00149700
CAS Number: 10031-53-5
EINECS: 233-380-3
UN: 1477
ADR: 5.1,II

Order Now

Specification Test Results

Appearance Semitransparent Crystalline solid
Total conc. of imp.as REO ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR