Aluminium nitrate nonahydrate 99+% for analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aluminum nitrate nonahydrate
AC21828-0025
AC21828-0100
AC21828-1000
AC21828-5000
TS21828-0025EA
221
USD
TS21828-0025
TS21828-0100
TS21828-1000
TS21828-5000
Aluminium nitrate nonahydrate 99+% for analysis
Aluminium nitrate nonahydrate
Formula:
Al(NO₃)₃·9H₂O MW: 375.13 g/mol Melting Pt: 73 Storage Temperature: Ambient |
MDL Number:
MFCD00149132 CAS Number: 7784-27-2 EINECS: 236-751-8 |
Specification Test Results
Appearance | Semitransparent to transparent Crystals |
Titration Complexometric | ≥99.0 % |
pH | 2.5 to 3.5 (5 % at 20°C) |
Chloride (Cl) | ≤20 ppm |
Sulfate (SO4) | ≤50 ppm |
Iron (Fe) | ≤20 ppm |
Heavy metals (as Pb) | ≤20 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...