Sodium dihydrogen phosphate dihydrate 98%, Extra Pure

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium phosphate monobasic dihydrateSodium dihydrogen orthophosphate dihydratePhosphoric acid (sodium hydrate)

AC44776-0050 AC44776-0010 AC44776-0250
TS44776-0010EA 58.3 USD
TS44776-0010 TS44776-0250 TS44776-0050
Sodium dihydrogen phosphate dihydrate 98%, Extra Pure
Sodium dihydrogen phosphate dihydrate
Formula: NaH₂PO₄·2H₂O
MW: 156.01 g/mol
Melting Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00149209
CAS Number: 13472-35-0

Order Now

Specification Test Results

Appearance Colorless to white Crystals
Loss on drying 21.5 % to 24.0 % (130°C)
pH 4.2 to 4.5 (5%, water)
Titration with NaOH 97.5 to 102.5 %
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR