Disodium hydrogen phosphate dihydrate ≥98%, Extra Pure

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium hydrogenphosphate dihydrateSorensen's saltsec-Sodium phosphatedi-Sodium hydrogen orthophosphate dihydrateDisodium phosphateSodium phosphate dibasic dihydratedi-Sodium hydrogen phosphate dihydrateSodium monohydrogen phosphate dihydrate

AC44815-0050 AC44815-0010
TS44815-0010EA 43.97 USD
TS44815-0010 TS44815-0050
Disodium hydrogen phosphate dihydrate ≥98%, Extra Pure
di-Sodium hydrogen phosphate dihydrate
Formula: Na₂HPO₄·2H₂O
MW: 177.99 g/mol
Melting Pt: 243…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00149182
CAS Number: 10028-24-7

Order Now

Specification Test Results

Appearance White to almost white or colorless Crystalline powder or crystals
Loss on drying 19.5 % to 21.0 % (130°C)
Solubility (5 % in water) Clear colorless
Titration with HClO4 97.5 to 102.5 % (dried substance)
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR